missing translation for 'onlineSavingsMsg'
Learn More

Multipette™ E3x Single Channel Pipet
GREENER_CHOICE

Provides high-precision dispensing regardless of the physical properties of the liquid. Eppendorf™ Multipette™ E3x Single Channel Pipet allows one-handed operation and contact free tip ejection.

£903.60 - £973.88

Specifications

Battery Type Lithium-ion
Compatible Tips Combitips advanced™, Eppendorf Quality™, PCR clean, Eppendorf Biopur™
Number of Channels 1
Type Liquid Dispenser
Volume (Metric) 1 to 50 mL
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15431920
GREENER_CHOICE
View Documents Promotion Details
Eppendorf
4987000029
Charging Adapter
£903.60
1 set
Please to purchase this item. Need a web account? Register with us today!
15471920
GREENER_CHOICE
View Documents Promotion Details
Eppendorf
4987000380
Charger Stand
£973.88
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Provides high-precision dispensing regardless of the physical properties of the liquid. Eppendorf™ Multipette™ E3x Single Channel Pipet allows one-handed operation and contact free tip ejection.

Creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
  • Full volume control, improved accuracy and reproducibility while reducing physical strain during use.
  • High speed dispensing with motorized piston
  • Automatic Combitip™ advanced™ tip recognition
  • One button tip ejection
  • Volume range from 1μl to 50mL
  • Li-ion battery
  • Illuminated display
  • Automatic dispensing
  • Aspirate (pipetting of supernatants)
  • Titrate
  • Sequential dispensing
  • Combined aspiration and dispensing mode
  • Includes 2 Combitips advanced™ assortment packs
Specifications

Specifications

Lithium-ion
1
1 to 50 mL
8
Combitips advanced™, Eppendorf Quality™, PCR clean, Eppendorf Biopur™
Liquid Dispenser
Illuminated color
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Multipette™ E3x Single Channel Pipet

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.