Learn More
Fisherbrand™ Aluminium Adapter with Wrench
Provides a stable base able to be extended at right angles to fit scaffolding feet. Fisherbrand™ Scaffolding Foot Adapters accept 3 x 13mm diameter rods or tubes at right angles - two perpendicular to the boss, the third in line with the boss.
Brand:  Fisherbrand™ 2119
Product Code. 11598112
  • £34.05 / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With Allen key and boss
  • With socket head screw fixings for rods or tubes
  • Manufactured from durable stainless steel
Note: For use with scaffolding feet (11578112).
Specifications
Specifications
Multiple Connector
For use with scaffolding feet 11578112.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Aluminium Adapter with Wrench > Product Type: Multiple Connector