missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-α-BOC-N(im)-tosyl-L-histidine, 98%, Thermo Scientific™
£35.32 - £124.08
Chemical Identifiers
CAS | 35899-43-5 |
---|---|
Molecular Formula | C18H23N3O6S |
Molecular Weight (g/mol) | 409.46 |
MDL Number | MFCD00065967 |
InChI Key | DCLJSEPKYJSEHW-UHFFFAOYNA-N |
Synonym | boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh |
PubChem CID | 6455010 |
IUPAC Name | (2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1 |
Description
Chemical Identifiers
35899-43-5 | |
409.46 | |
DCLJSEPKYJSEHW-UHFFFAOYNA-N | |
6455010 | |
CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1 |
C18H23N3O6S | |
MFCD00065967 | |
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh | |
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
35899-43-5 | |
97.5% min. (HPLC) | |
MFCD00065967 | |
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh | |
CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1 | |
409.46 | |
409.46 | |
N-α-BOC-N(im)-tosyl-L-histidine |
117.0°C to 118.0°C | |
C18H23N3O6S | |
+ 17.30 | |
DCLJSEPKYJSEHW-UHFFFAOYNA-N | |
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
6455010 | |
98% |
RUO – Research Use Only