missing translation for 'onlineSavingsMsg'
Learn More
N-Benzyl-4-piperidone, 99%, Thermo Scientific Chemicals Product Code.: 10723831

N-Benzyl-4-piperidone, 99%, Thermo Scientific Chemicals

Product Code. 10723831
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10723831

Brand: Thermo Scientific Acros 106131000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.26
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-oneShow MoreShow Less
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Straw
Density 1.0600g/mL
Boiling Point 133.0°C to 135.0°C (7.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410
Quantity 100 mL
Beilstein 21,IV,3189
Specific Gravity 1.06
Solubility Information Solubility in water: 12g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, d.m.f.,, acetone and chloroform
Formula Weight 189.26
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material N-Benzyl-4-piperidone, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462378 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number106132500,106131000
Lot NumberA0462378 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number106132500,106131000
Lot NumberA0438495 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number106132500,106131000
Lot NumberA0408051 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number106132500,106131000
Lot NumberA0357913 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number106132500,106131000
compliance-icons
Product Identifier
  • N-Benzyl-4-piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyl-4-piperidone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.