Learn More
N-Benzylglycine ethyl ester, 97%, Thermo Scientific Chemicals
Description
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Specifications
Specifications
Boiling Point | 102°C to 104°C (1mmHg) |
Refractive Index | 1.5053 |
Beilstein | 2104816 |
Solubility Information | Not miscible or difficult to mix in water. |
Density | 1.048 |
Quantity | 500 g |
Flash Point | 112°C (233°F) |
Formula Weight | 193.25 |
Percent Purity | 97% |
Chemical Name or Material | N-Benzylglycine ethyl ester |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W23K004 | Certificate of Analysis | 05/10/2024 | L15496.22,L15496.14,L15496.36 |
W05J031 | Certificate of Analysis | 21/09/2023 | L15496.22,L15496.14,L15496.36 |
10149914 | Certificate of Analysis | 07/12/2021 | L15496.22,L15496.14,L15496.36 |
FA023291 | Certificate of Analysis | 06/12/2021 | L15496.22,L15496.14,L15496.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.