missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals Product Code.: 15448757

N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15448757
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15448757

Brand: Thermo Scientific Alfa Aesar H52191.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254101-10-5
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD01862861
InChI Key FAFJSSKTLCNWRJ-CYBMUJFWSA-N
Synonym boc-beta-hoasp obzl-oh, boc-l-beta-glutamic acid 5-benzyl ester, boc-beta-glu obzl-oh, boc-, a-hoasp obzl-oh, r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid, 3r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid, fmoc-asparaginol, boc-beta-homoaspartic acid obzl, ambotzbaa6150, boc-hoasp obzl-ohShow MoreShow Less
PubChem CID 2761513
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)CC(=O)OCC1=CC=CC=C1

Specifications

Beilstein 8430789
Quantity 250 mg
Formula Weight 337.37
Percent Purity 95%
Chemical Name or Material N-Boc-L-beta-glutamic acid 5-benzyl ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK19W037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52191.03,H52191.MD
Lot NumberF13Z020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52191.03,H52191.MD
Lot NumberP03H031 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52191.03,H52191.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.