missing translation for 'onlineSavingsMsg'
Learn More
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals Product Code.: 11480078

n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals

Product Code. 11480078
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11480078

Brand: Thermo Scientific Alfa Aesar B23673.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 3 g/L (20°C). Soluble in chloroform and methanol.

Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-34-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009454
InChI Key UZKWTJUDCOPSNM-UHFFFAOYSA-N
Synonym n-butyl vinyl ether, butyl vinyl ether, vinyl butyl ether, butane, 1-ethenyloxy, butoxyethylene, 1-ethenyloxy butane, butoxyethene, ether, butyl vinyl, vinyl n-butyl ether, ethenyl n-butyl etherShow MoreShow Less
PubChem CID 8108
IUPAC Name 1-ethenoxybutane
SMILES CCCCOC=C

Specifications

Melting Point -113°C
Density 0.778
Boiling Point 92°C to 94°C
Flash Point −12°C (10°F)
Linear Formula CH3(CH2)3OCH=CH2
Refractive Index 1.401
Quantity 100 mL
UN Number UN3271
Beilstein 1560217
Solubility Information Soluble in water 3g/L (20°C). Soluble in chloroform and methanol.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material n-Butyl vinyl ether, stabilized with 0.01% KOH
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253817 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberB23673.AE,B23673.AP
Lot Number10237275 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB23673.AE,B23673.AP
Lot Number10230956 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23673.AE,B23673.AP
Lot Number5013R05M Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23673.AE,B23673.AP
Lot NumberI5226A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23673.AE,B23673.AP
compliance-icons compliance-icons
Product Identifier
  • n-Butyl vinyl ether
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Butyl vinyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.