missing translation for 'onlineSavingsMsg'
Learn More
n-Dodecane, 99%, pure, Thermo Scientific Chemicals Product Code.: 10221463

n-Dodecane, 99%, pure, Thermo Scientific Chemicals

Product Code. 10221463
250 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
250 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10221463

Brand: Thermo Scientific Acros 117592500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661Show MoreShow Less
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -12.0°C
Density 0.7530g/mL
Boiling Point 215.0°C to 217.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H26
Linear Formula CH3(CH2)10CH3
Refractive Index 1.4210 to 1.4230
Quantity 250 mL
Beilstein 01, 171
Specific Gravity 0.753
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Viscosity <7 mm2/s (40°C)
Formula Weight 170.34
Vapor Pressure 1mbar at 48°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Dodecane, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466724 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number117595000,117590100,117590025,117592500,117590250,117591000
Lot NumberA0460544 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number117595000,117590100,117590025,117592500,117590250,117591000
Lot NumberB0542455A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number117595000,117590100,117590025,117592500,117590250,117591000
Lot NumberB0541412A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number117595000,117590100,117590025,117592500,117590250,117591000
Lot NumberA0429668 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number117595000,117590100,117590025,117592500,117590250,117591000
compliance-icons
Product Identifier
  • Dodecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Dodecane, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.