Learn More
Chemical Identifiers
CAS | 112-40-3 |
---|---|
Molecular Weight (g/mol) | 170.34 |
MDL Number | MFCD00008969 |
InChI Key | SNRUBQQJIBEYMU-UHFFFAOYSA-N |
Synonym | n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 |
PubChem CID | 8182 |
ChEBI | CHEBI:28817 |
IUPAC Name | dodecane |
SMILES | CCCCCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10068531
|
Thermo Scientific Acros
117590250 |
25 mL | Glass bottle |
£18.79
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10274620
|
Thermo Scientific Acros
117591000 |
100 mL | Glass bottle |
£36.88
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10221463
|
Thermo Scientific Acros
117592500 |
250 mL | Glass bottle |
£68.74
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645152
|
Thermo Scientific Acros
117595000 |
500 mL | Glass Bottle |
£107.54
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247433
|
Thermo Scientific Acros
117590025 |
2.5 L | Glass bottle |
£337.68
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15264604
|
Thermo Scientific Acros
117590100 |
10 L | Metal Drum |
£990.93
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-40-3 | |
MFCD00008969 | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
CHEBI:28817 | |
CCCCCCCCCCCC |
170.34 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
8182 | |
dodecane |
Specifications
112-40-3 | |
0.7530g/mL | |
71°C | |
99% | |
CH3(CH2)10CH3 | |
MFCD00008969 | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
dodecane | |
8182 | |
170.34 | |
99% | |
Liquid |
-12.0°C | |
215.0°C to 217.0°C | |
Authentic | |
C12H26 | |
1.4210 to 1.4230 | |
01, 171 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCCCCCCCCCC | |
170.34 | |
CHEBI:28817 | |
1mbar at 48°C | |
Pure | |
n-Dodecane, 99% |
Safety and Handling
GHS P Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Combustible liquid. Harmful by inhalation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking.
NFPA
Danger
missing translation for 'einecsNumber' : 203-967-9
missing translation for 'rtecsNumber' : JR2125000
missing translation for 'tsca' : TSCA
- n-Dodecane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only