missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™

High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis

£89.00 - £230.00

Chemical Identifiers

CAS 110-54-3, 107-83-5, 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
16594334
View Documents Promotion Details
Fisher Chemical
H306-1
1 L Amber Soda Lime Glass
£89.00
1 L
Please to purchase this item. Need a web account? Register with us today!
16504344
View Documents Promotion Details
Fisher Chemical
H306-4
4 L Amber Soda Lime Glass
£230.00
4 L
Please to purchase this item. Need a web account? Register with us today!
16514344
View Documents Promotion Details
Fisher Chemical
H306SK-4
4 L Safe-Cote™ Amber Glass 
£230.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Chemical Identifiers

110-54-3, 107-83-5, 96-14-0
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-95°C
110-54-3, 107-83-5, 96-14-0
0.659g/cm³
Pass Test
1.3710 to 1.3750 (at 25°C)
1ppm max.
MFCD02179311
1208
0.01% max.
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.17
n-Hexane
0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.07 AU max. at 220nm, 0.25 AU max. at 210nm, 1.00 AU max. at 195nm
Colorless
69°C
Liquid
95%
C6H14
160mbar at 20°C
0.31 mPa/s at 20°C
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.18
CHEBI:29021
Also meets ACS specifications
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid

Product Identifier
  • n-Hexane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : hexane, 2-methylpentane, 3-methylpentane

SDS
Documents

Documents

Product Certifications