missing translation for 'onlineSavingsMsg'
Learn More
N'-Hydroxy-3-methyl-2-pyridinecarboximidamide, 97%, Thermo Scientific™ Product Code.: 10738603

N'-Hydroxy-3-methyl-2-pyridinecarboximidamide, 97%, Thermo Scientific™

Product Code. 10738603
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10738603

Brand: Thermo Scientific Maybridge MO01289DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-33-8
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD06200877
InChI Key YIBWTWSINGSDFJ-UHFFFAOYSA-N
Synonym 3-methylpyridine-2-amidoxime, n'-hydroxy-3-methyl-2-pyridinecarboximidamide, n'-hydroxy-3-methylpyridine-2-carboximidamide, z-n'-hydroxy-3-methylpyridine-2-carboximidamide, uqdfgzccscdnso-uhfffaoysa-n, n-hydroxy-3-methyl-pyridine-2-carboxamidine, 3-methyl-1h-pyridin-2-ylidene-nitrosomethanamine, 3-methyl-1h-pyridin-2-ylidene nitroso methanamineShow MoreShow Less
PubChem CID 6816486
IUPAC Name (3-methyl-1H-pyridin-2-ylidene)-nitrosomethanamine
SMILES CC1=CC=CNC1=C(N)N=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material N'-Hydroxy-3-methyl-2-pyridinecarboximidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N'-Hydroxy-3-methyl-2-pyridinecarboximidamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.