Learn More
N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals
N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2
£22.41 - £202.01
Chemical Identifiers
CAS | 6283-63-2 |
---|---|
Molecular Formula | C10H16N2·H2SO4 |
Molecular Weight (g/mol) | 262.33 |
MDL Number | MFCD00012993 |
InChI Key | AYLDJQABCMPYEN-UHFFFAOYSA-N |
Synonym | n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1 |
PubChem CID | 80166 |
IUPAC Name | 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid |
SMILES | CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10197133
|
Thermo Scientific Acros
395000500 |
50 g | Glass bottle |
£22.41
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157183
|
Thermo Scientific Acros
395002500 |
250 g | Glass bottle |
£68.04
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10085053
|
Thermo Scientific Acros
395000010 |
1 kg | Glass bottle |
£202.01
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6283-63-2 | |
262.33 | |
AYLDJQABCMPYEN-UHFFFAOYSA-N | |
80166 | |
CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O |
C10H16N2·H2SO4 | |
MFCD00012993 | |
n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1 | |
4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid |
Specifications
6283-63-2 | |
100.0 | |
White to Beige | |
99% | |
MFCD00012993 | |
AYLDJQABCMPYEN-UHFFFAOYSA-N | |
4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid | |
80166 | |
99% | |
N, N-Diethyl-p-phenylenediamine sulfate |
98.5 | |
184.0°C to 187.0°C | |
Authentic | |
C10H16N2·H2SO4 | |
n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1 | |
CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O | |
262.33 | |
262.33 | |
Crystalline Powder or Crystals With Lumps |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-500-6
- N,N-Diethyl-p-phenylenediamine sulfate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only