missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10154440

N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10154440
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
This item is not returnable. View return policy

Product Code. 10154440

Brand: Thermo Scientific Acros 181070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow MoreShow Less
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 0.9370g/mL
Quantity 1 L
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Assay Percent Range 99+%
Residue after Evaporation 0.01% max.
Linear Formula CH3CON(CH3)2
Vapor Pressure Vapor Pressure: 1.7mbar at 25°C
Beilstein 04, 59
Water 0.1% max.
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Formula Weight 87.12
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453545 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number181070010,181070025,181070250,181070050
Lot NumberA0467265 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number181070010,181070025,181070250,181070050
Lot NumberA0463426 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number181070010,181070025,181070250,181070050
Lot NumberA0459489 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number181070010,181070025,181070250,181070050
Lot NumberB0542397B Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number181070010,181070025,181070250,181070050
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylacetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.