missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylacetamide, 99%, Reagent Grade, Honeywell™ Product Code.: 15637790

N,N-Dimethylacetamide, 99%, Reagent Grade, Honeywell™

Product Code. 15637790 Shop All Honeywell Research Chemicals Products
2.5 L, Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy

Product Code. 15637790

Brand: Honeywell Research Chemicals D1375102.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow MoreShow Less
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

pH 4 (at 200.00g/L 20°C)
Melting Point -20°C
Color Colorless to Yellow
Density 0.937g/mL at 25°C
Boiling Point 164°C to 166°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 11.5% (160°F), 1.8% (100°F)
Flash Point 70°C (158°F)
Ignition Point 914°F
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CON(CH3)2
Vapor Pressure 4mmHg (38°C), 2mmHg (25°C)
Beilstein 1737614
Solubility Information Fully miscible in water
Formula Weight 87.12g/mol
Percent Purity 0.99
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylacetamide, 99%, Reagent Grade, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.