missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals Product Code.: 11368177

N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Product Code. 11368177
1 L
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11368177

Brand: Thermo Scientific Alfa Aesar 022916.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow MoreShow Less
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Color Colorless
Appearance Colorless liquid
Density 0.940 g/cm3 at 20°C
Boiling Point 165°C to 166°C
Quantity 1 L
Flash Point 70°C (158°F)
Physical Form Liquid
Refractive Index 1.4385
Assay Percent Range >99.5%
Vapor Pressure 3.3 hPa (3mm Hg) at 20°C
Beilstein 1737614
Solubility Information Miscible with water,benzene,alcohol,acetone,ether,esters and ketones.
Formula Weight 87.12
Grade HPLC
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443842 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog Number022916.K2,022916.0F,022916.M1,022916.K7
Lot NumberA0454462 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number022916.K2,022916.0F,022916.M1,022916.K7
Lot NumberN26G704 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number022916.K2,022916.M1,022916.K7,022916
Lot NumberJ30R844 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number022916.K2,022916.0F,022916.M1,022920.M1,022916.K7
Lot NumberN28I751 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog Number022916.K2,022916.0F,022916.M1,022920.M1,022916.K7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.