Learn More
n-Tetradecane, 99+%, Thermo Scientific Chemicals
Description
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 6°C |
Density | 0.763 g/mL |
Boiling Point | 252°C to 254°C |
Flash Point | 99°C (211°F) |
Assay Percent Range | >99% |
Refractive Index | 1.429 |
Quantity | 25 g |
Solubility Information | Soluble in ether,acetone and alcohol. Insoluble in water. |
Formula Weight | 198.39 |
Physical Form | Liquid |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S06I042 | Certificate of Analysis | 17/11/2022 | 030809.22,030809.14,030809.36 |
S02I029 | Certificate of Analysis | 09/06/2022 | 030809.22,030809.14,030809.36 |
R05I043 | Certificate of Analysis | 24/05/2022 | 030809.22,030809.14,030809.36 |
Y09H039 | Certificate of Analysis | 12/04/2022 | 030809.22,030809.14,030809.36 |
M10I025 | Certificate of Analysis | 12/04/2022 | 030809.22,030809.14,030809.36 |
Safety and Handling

- n-Tetradecane
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contains : Tetradecane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.