missing translation for 'onlineSavingsMsg'
Learn More
n-Undecane, 99%, Thermo Scientific Chemicals Product Code.: 11318297

n-Undecane, 99%, Thermo Scientific Chemicals

Product Code. 11318297
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11318297

Brand: Thermo Scientific Alfa Aesar 031807.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, undShow MoreShow Less
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

Melting Point -26°C
Color Colorless
Density 0.74 g/mL
Boiling Point 196°C
Flash Point 60°C (140°F)
Odor Mild
Linear Formula CH3(CH2)9CH3
Refractive Index 1.4173
Quantity 250 g
UN Number UN2330
Solubility Information Miscible with ethyl alcohol and ether. Immiscible with water.
Formula Weight 156.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Undecane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY06K020 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog Number031807.30,031807.18
Lot NumberT16G049 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number031807.30,031807.18
Lot NumberZ15J045 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number031807.30,031807.18
Lot NumberW09I030 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number031807.30,031807.18
Lot NumberY09H040 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number031807.30,031807.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Undecane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.