missing translation for 'onlineSavingsMsg'
Learn More
N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™ Product Code.: 10079441

N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. 10079441
1 g
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 10079441

Brand: Thermo Scientific Maybridge TL00405DA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 41419-59-4
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00052332
InChI Key MGCCWCLGIPNIBP-UHFFFAOYSA-N
Synonym n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amineShow MoreShow Less
PubChem CID 737351
IUPAC Name N-methyl-4-(trifluoromethoxy)aniline
SMILES CNC1=CC=C(C=C1)OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NHCH3
Quantity 1 g
Formula Weight 191.15
Percent Purity 95%
Chemical Name or Material N1-methyl-4-(trifluoromethoxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.