Learn More
Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes
Enhance aseptic techniques with these externally threaded general long-term storage cryogenic tubes, ideal for research and general lab storage.
£22.85 - £679.00
Specifications
Product Type | Cryogenic Screw Top |
---|---|
Material | Polypropylene |
Thread Style | External |
Product Line | Nalgene™ |
Alert:
Do not store in the liquid phase of nitrogen without CryoFlex tubing
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
11375644
|
Thermo Scientific™
5000-1012 |
1.0 mL | Sterile |
£679.00
Pack of 500 |
In Stock
Log in to see stock. |
||||
11740573
|
Thermo Scientific™
5000-0012 |
1.2 mL | Sterile |
£429.00
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11385644
|
Thermo Scientific™
5000-1020 |
1.5 mL | Sterile |
£659.00
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11750573
|
Thermo Scientific™
5000-0020 |
2.0 mL | Sterile |
£447.00
Pack of 500 |
In Stock
Log in to see stock. |
||||
10344691
|
Thermo Scientific™
5000-0020 |
2.0 mL | Sterile |
£22.85
Pack of 25 |
In Stock
Log in to see stock. |
||||
11365644
|
Thermo Scientific™
5000-0050 |
5.0 mL | Sterile |
£290.00
Pack of 250 |
In Stock
Log in to see stock. |
||||
10664321
|
Thermo Scientific™
5005-0015 |
15 mL | Sterile |
£201.00
Pack of 75 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10199850
|
Thermo Scientific™
5011-0012 |
1.2 mL | Non-sterile |
£379.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543611
|
Thermo Scientific™
5011-0020 |
2.0 mL | Non-sterile |
£394.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10662102
|
Thermo Scientific™
5012-0012 |
1.2 mL | Sterile |
£485.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731432
|
Thermo Scientific™
5012-0020 |
2.0 mL | Sterile |
£506.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Enhance aseptic techniques with externally threaded Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes. These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to vapor phase liquid nitrogen temperatures.
- Large writing areas accommodate more written sample information and graduations for easy visual volume assessment
- Noncytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
- Certified SAL of 10-6 for the highest sterility assurance available
- Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
- System 100 Tubes offer a more dense storage option (10x10) to save precious storage space
- Self standing tubes for ease of use on the benchtop
- Blank, nonsterile and uncapped tubes available in bulk for less demanding applications
Especificaciones
Cryogenic Screw Top | |
Polypropylene | |
External | |
Nalgene™ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.