Learn More
Thermo Scientific™ Nalgene™ Large HDPE Funnels

Choose a safer alternative to glass with this shatterproof funnel, featuring external and internal ribbing for air release.
£51.05 - £113.00
Product Code | Brand | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity & Availability | |||||
10788681
![]() |
Thermo Scientific™
4262-0080 |
201mm | 1.9 L |
£51.05
Each |
In Stock
Log in to see stock. ![]() |
|||||
10122591
![]() |
Thermo Scientific™
4262-0110 |
258mm | 4.06 L |
£113.00
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
- High-density polyethylene is shatterproof and dentproof, making it a reliable long-term tool for the lab
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling High Density Polyethylene (HDPE) labware at end of product life. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
HDPE | |
Not Autoclavable | |
1/Pk.; 6/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.