missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure Product Code.: 10420391
In Stock
to see stock.
Add to basket

Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure
GREENER_CHOICE

Product Code. 10420391
30 mL, 20 mm
Click to view available options
Capacity (Metric):
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20 mm
24 mm
28 mm
38-430 mm

Product Code. 10420391

Brand: Thermo Scientific™ 20030001

In Stock
to see stock.
Edge
Add to basket
Add to basket

Carry out applications such as trace metal analysis with flexible, impact- and chemical-resistant LDPE bottles with closure.

Carry out applications such as trace metal analysis with flexible, impact- and chemical-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure, which are extremely low in trace metal content. The bottles are leakproof† when used with Nalgene closures and provide excellent visibility of contents, as well as excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:

  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Translucence allows easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof† protection without use of liner that can wrinkle, cause leaks or contaminate your reagents

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Natural
Description Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 14 mm
Material LDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural
Neck Finish 20 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 61 mm
Product Type Narrow Mouth Bottle
Width (English) 1.35 in.
Height (English) 2.4 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 20 mm
Diameter (Metric) 34 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424972 Certificate TypeCertificate of Compliance Date04/04/2025 Catalog Number2003-0001
Lot Number1415152 Certificate TypeCertificate of Compliance Date12/10/2024 Catalog Number2003-0001
Lot Number1409009 Certificate TypeCertificate of Compliance Date03/07/2024 Catalog Number2003-0001
Lot Number1402081 Certificate TypeCertificate of Compliance Date08/03/2024 Catalog Number2003-0001
Lot Number1392733 Certificate TypeCertificate of Compliance Date14/10/2023 Catalog Number2003-0001

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.