Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks
GREENER_CHOICE
Increase mixing efficiency with these polycarbonate flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae.
£59.97 - £194.00
Specifications
Material Polycarbonate
Autoclavable Autoclavable
Graduated Yes
Bottom Shape Baffled
View More Specs
Alert:
Repeated autoclaving may result in some loss of mechanical strength.
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10768911
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4110-0250
250 mL
£59.97
Pack of 4
In Stock
to see stock.
Add to basket
10190441
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4110-0500
500 mL
£71.40
Pack of 4
In Stock
to see stock.
Add to basket
10607341
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4110-1000
1000 mL
£63.75
Pack of 2
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10094900
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4110-2000
2000 mL
£194.00
Pack of 2
In Stock
to see stock.
Edge
Add to basket
Description
Description
Increase mixing efficiency using Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
  • Polycarbonate is break-resistant and clear; easy to read graduations help identify liquid level with ease.
  • Bottom baffles provide increased mixing, aeration, and gas exchange.
  • Flasks are excellent for a variety of lab procedures, including fermentation, cell culture grown and harvesting, and generic engineering procedures.
  • 500 and 100mL sizes accept either standard 38mm stainless-steel or plastic closures.
  • Flasks are autoclavable.
Note: Repeated autoclaving may result in some loss of mechanical strength.  Wash and rinse well before autoclaving.
Specifications
Specifications
Polycarbonate
Autoclavable
Yes
Baffled
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks