Learn More
Thermo Scientific™ Nalgene™ Polycarbonate, Carboy with Sanitary Neck
Description
Clean the one-piece molded, non-threaded Thermo Scientific™ Nalgene™ Polycarbonate Sanitary Carboy more easily than threaded containers. Designed for use as receiver or dispensing vessel in biopharmaceutical applications, the 3 in. sanitary flange molded on the neck accepts standard flange fittings. A clamping closure system securely seals and will not back off.
- Solid ported sanitary end caps are available
- Made of same polycarbonate (PC) resin as Nalgene PC Clearboys and PC Bottles, permitting switch to Sanitary PC Carboys without material validation issues
- To seal closure, use with Nalgene Heavy Duty Clamp (Cat. No. 2865-0300) and Gasket (Cat. No. 2672-0300)
- Autoclavable

Specifications
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Size | 3 in. |
Color | Clear |
Description | Polycarbonate Sanitary Carboy |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Product Information
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1422527 | Certificate of Compliance | 04/02/2025 | 2261-0050 |
1415212 | Certificate of Compliance | 29/10/2024 | 2261-0050 |
1398475 | Certificate of Compliance | 05/01/2024 | 2261-0050 |
1319957 | Certificate of Compliance | 02/06/2021 | 2261-0050 |
1267092 | Certificate of Compliance | 16/10/2019 | 2261-0050 |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.