Learn More
Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
GREENER_CHOICE
Ensure contamination-free dispensing with these chemical-resistant wash bottles made with Teflon™ fluoropolymer.
£154.00 - £552.00
Specifications
Color Translucent
Material FEP
Closure Included Yes
Autoclavable Autoclavable
Graduated No
View More Specs
Products 4
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10508131
GREENER_CHOICE
View Documents
Thermo Scientific™
2403-0125
125 mL 24 mm
£154.00
Each
In Stock
to see stock.
Add to basket
10548701
GREENER_CHOICE
View Documents
Thermo Scientific™
2403-0250
250 mL 24 mm
£266.00
Each
In Stock
to see stock.
Add to basket
10170551
GREENER_CHOICE
View Documents
Thermo Scientific™
2403-0500
500 mL 38-430 mm
£220.00
Each
In Stock
to see stock.
Add to basket
15951491
GREENER_CHOICE
View Documents
Thermo Scientific™
2403-1000
1000 mL 38-430 mm
£552.00
Pack of 2
In Stock
to see stock.
Add to basket
Description
Description
Ensure contamination-free dispensing with Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer, useful for trace element work and other high-purity applications. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymer. Designed to dispense through the top with a closure and stem molded in one piece for reliable service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
  • With ETFE screw closure/stem and draw tube
  • Demonstrates excellent chemical resistance to nearly any chemical in your lab including solvents—won't become brittle or crack
  • Inert fluoropolymer construction is low in extractables and low binding—useful for trace element work
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Autoclavable/Transparent
  • Sputter-free, precision stream
.
Specifications
Specifications
Translucent
Yes
No
Cylindrical
ETFE
Yes
FEP
Autoclavable
Narrow
Wash Bottle
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.