missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures

Choose these torque wrench fittings designed for specific HDPE closures to achieve accurate torque readings when applying closures onto bottles.

Brand:  Thermo Scientific™ 2195-1021

 View more versions of this product

Product Code. 11369754

This item is not returnable. View return policy

Description

Description

Choose Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures to achieve accurate torque readings when applying closures onto bottles. Proper torquing of Nalgene closures is critical to leakproof† sealing.

  • Epoxy
  • Specifically designed for Nalgene HDPE closures

Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fittings
1/4 in.
Epoxy
20 to 415 for serum vial
Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177), and Nalgene Serum Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7230030010 Certificate TypeCertificate of Compliance Date19/07/2017 Catalog Number2195-1020,2195-1021
Lot Number7241729020-COC Certificate TypeCertificate of Compliance Date14/04/2017 Catalog Number2195-1020,2195-1021
Lot Number7152889010-COC Certificate TypeCertificate of Compliance Date12/04/2017 Catalog Number2195-1020,2195-1021
Lot Number7139503010-COC Certificate TypeCertificate of Compliance Date12/04/2017 Catalog Number2195-1020,2195-1021
Lot Number7248195040-COC Certificate TypeDeclaration of Conformity Date25/03/2017 Catalog Number2195-1020,2195-1021
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures > Drive size: 1/4 in.; Closure size: 20-415 for serum vial closure

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.