missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Unwire™ Half-Racks
GREENER_CHOICE

Space-efficient half-racks are ideal for use in incubators, refrigerators, freezers and under fume hoods.

£33.45 - £40.45

Specifications

Product Type Test Tube Half Rack
Autoclavable Autoclavable
Positions 36
View More Specs

Products 4
Product Code Brand Color Holds Price Quantity & Availability  
Product Code Brand Color Holds Price Quantity & Availability  
10170591
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5972-0013
White Tubes (13 mm)
£34.15
Each
In Stock
to see stock.
Add to basket
 
10633911
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5972-0313
Blue Tubes (13 mm)
£33.45
Each
In Stock
to see stock.
Add to basket
 
10549461
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5972-0016
White Tubes (16 mm)
£40.45
Each
In Stock
to see stock.
Add to basket
 
10537541
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5972-0316
Blue Tubes (16 mm)
£39.85
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Fit easily in ice or water baths without floating.

  • Made of durable ResMere™ performance plastic
  • Withstand most lab chemicals and temperatures up to 121°C (249.8°F)
  • Repeatedly autoclavable
  • Each rack holds 36 tubes

Specifications

Specifications

Test Tube Half Rack
Autoclavable
36
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Unwire™ Half-Racks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.