missing translation for 'onlineSavingsMsg'
Learn More
Nalidixic acid, 99.5%, Thermo Scientific Chemicals Product Code.: 10711831

Nalidixic acid, 99.5%, Thermo Scientific Chemicals

Product Code. 10711831
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10711831

Brand: Thermo Scientific Acros 169900250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nalixidic acid, or nalixidin, CAS # 389-08-2, is a quinolone antibiotic used to inhibit DNA gyrase and prevent cell division.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalixidic acid is a synthetic quinolone that is a monocarboxylic acid composed of 1,8-naphthyridine substituted with carboxylic acid, ethyl, and methyl groups

• Nalixidic acid is a DNA gyrase inhibitor that prevents DNA synthesis and subsequent cell divisions

Applications

• Nalixidic acid is primarily effective against gram-negative bacteria

• It may be useful for bacterial resistance studies and laboratory research on drug allergies and cross-reactivity

• Nalixidic acid is suitable for high-performance liquid chromatography (HPLC) and related biochemistry laboratory applications

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.24
MDL Number MFCD00006884
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 225.0°C to 231.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass bottle
Quantity 25 g
Beilstein 25, V,7, 384
Merck Index 15, 6444
Solubility Information Solubility in water: 0.1g/L (23°C). Other solubilities: soluble in alkali solutions,35mg/mL chloroform- 1.6mg/mL toluene (23°C),1.3mg/mL methanol- 0.9mg/mL ethanol (23°C),0.1mg/mL ether (23°C)
Formula Weight 232.24
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material Nalidixic acid, 99.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465945 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number169901000,169900250,169900050
Lot NumberA0461932 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number169901000,169900250,169900050
Lot NumberA0432661 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number169901000,169900250,169900050
Lot NumberA0446177 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number169901000,169900250,169900050
Lot NumberA0415644 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog Number169901000,169900250,169900050
compliance-icons compliance-icons
Product Identifier
  • Nalidixic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nalidixic acid, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.