missing translation for 'onlineSavingsMsg'
Learn More
Naltrexone hydrochloride, Tocris Bioscience™ Product Code.: 18782724

Naltrexone hydrochloride, Tocris Bioscience™

Product Code. 18782724
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 18782724

Brand: Tocris Bioscience 0677/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective opioid antagonist

Opioid antagonist.
TRUSTED_SUSTAINABILITY

Specifications

CAS 16676-29-2
Molecular Formula C20H24ClNO4
Quantity 100 mg
Synonym naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin
InChI Key RHBRMCOKKKZVRY-ITLPAZOVSA-N
SMILES C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
Molecular Weight (g/mol) 377.865
PubChem CID 5485201
Formula Weight 377.87
Percent Purity >99%
Chemical Name or Material Naltrexone hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Naltrexone hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.