Learn More
Thermo Scientific NanoDrop™ Reagents
Certified standards for verifying instrument performance to factory specifications are available separately.
Brand:  Thermo Scientific CHEM-PV-8
 View more versions of this product
Product Code. 30089480
  • £157.00 / Each
Explore more special offers
Description
Description
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with NanoDrop Eight UV-Vis Spectrophotometer Solution with certified absorbance values. Use to create a record of correct photometric performance.
Contains: Two vials of CHEM-PV-1, two pastuer pipettes, and two 8-well PCR strip tubes.
Intended for single use only.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NanoDrop PV-8 Performance Verification Kit
NanoDrop Eight
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific NanoDrop™ Reagents >