Learn More
Chemical Identifiers
CAS | 91-20-3 |
---|---|
Molecular Formula | C10H8 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00001742 |
InChI Key | UFWIBTONFRDIAS-UHFFFAOYSA-N |
Synonym | naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene |
PubChem CID | 931 |
ChEBI | CHEBI:16482 |
IUPAC Name | naphthalene |
SMILES | C1=CC2=CC=CC=C2C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10153910
|
Thermo Scientific Acros
180902500 |
250 g | Plastic bottle |
£17.29
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337650
|
Thermo Scientific Acros
180900010 |
1 kg | Plastic bottle |
£35.78
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10287023
|
Thermo Scientific Acros
180900025 |
2.5 kg | Plastic bottle |
£74.07
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13478418
|
Thermo Scientific Acros
180900050 |
5 kg | Plastic drum |
£112.56
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-20-3 | |
128.17 | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
931 | |
naphthalene |
C10H8 | |
MFCD00001742 | |
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene | |
CHEBI:16482 | |
C1=CC2=CC=CC=C2C=C1 |
Specifications
91-20-3 | |
100.0 | |
White | |
218.0°C | |
Authentic | |
C10H8 | |
MFCD00001742 | |
15,6455 | |
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
naphthalene | |
931 | |
128.17 | |
Crystalline Flakes, Crystals, Tablets, Balls or Crystalline Powder |
98.5 | |
79.0°C to 82.0°C | |
0.9900g/mL | |
78°C | |
98.5% min. (GC) | |
C10H8 | |
05,IV,164 | |
0.99 | |
Solubility in water: 30mg/l (25°C). Other solubilities: soluble in ethanol,,very soluble in acetone,chloroform,ether,,benzene,toluene,carbon disulfide | |
C1=CC2=CC=CC=C2C=C1 | |
128.17 | |
CHEBI:16482 | |
99% | |
Naphthalene |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Flammable solid.
GHS P Statement
Obtain special instructions before use.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smokin
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-049-5
- Naphthalene
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.