Learn More
Whirl-Pak™ Write-On Scoop Sterile Sample Bag
GREENER_CHOICE
Sterile scoop already contained inside the bag eliminates the need to carry two items to collect a sample and minimizes contamination.
Brand:  Whirl-Pak™ B01350WA
Product Code. 16499930
  • £149.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Sterile scoop is already contained inside the bag, eliminating the need to carry two items to collect the sample and minimizing contamination. The 2 oz. (60 cc) polypropylene scoop measures 7 in. (18.5 cm) long.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate, bags should not be used at temperatures above 180°F (82°C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346°F (-210°C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Whirl-Pak™ Write-On Scoop Sterile Sample Bag > Capacity: 18 oz. bag with 2 oz. scoop