Learn More
Abeba™ Nature 4055 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 4055/46
 View more versions of this product
Product Code. 15691368
  • £81.20 / 1 pair
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
  • Resistant to oil and petrol
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
46
Rubber
Black
Leather
Slip-resistant
4055
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ Nature 4055 Shoes > Size: 46