Learn More
Cytiva Ni Sepharose™ 6 Fast Flow
Combines the advantages of Ni2 for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform. Cytiva Lifescience™ Ni Sepharose™ 6 Fast Flow is part of a range of IMAC chromatography media for capture and intermediate purification.
Brand:  Cytiva 17-5318-02
 View more versions of this product
Product Code. 10343060
  • £1178.00 / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.
  • Designed for scaling up purification of histidine-tagged proteins.
  • High protein binding capacity and minimal leakage of Ni2+ ions.
  • As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.
  • Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
  • The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1M HCl, 0.1M NaOH, 8M Urea
100 mL
250mm
1 bar
90μm
4° to 30°C
Ni Sepharose 6 FF chromatography media
9.84 in.
600 cm/hr.
3 to 12
6% Cross-linked Agarose
Ni-Sepharose
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3