missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ni Sepharose™ 6 Fast Flow
Combines the advantages of Ni2 for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform. Cytiva Lifescience™ Ni Sepharose™ 6 Fast Flow is part of a range of IMAC chromatography media for capture and intermediate purification.
£78.43 - £4808.00
Specifications
Buffer | 0.1M HCl, 0.1M NaOH, 8M Urea |
---|---|
Product Type | Ni Sepharose 6 FF chromatography media |
Height (English) | 9.84 in. |
Height (Metric) | 250mm |
Flow Rate | 600cm/hr. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11540814
|
Cytiva
17-5318-06 |
5mL |
£78.43
5mL |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
10291210
|
Cytiva
17-5318-01 |
25mL |
£327.00
25mL |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10343060
|
Cytiva
17-5318-02 |
100mL |
£1145.00
100mL |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
10353060
|
Cytiva
17-5318-03 |
500mL |
£4808.00
500mL |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
Description
Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.
- Designed for scaling up purification of histidine-tagged proteins.
- High protein binding capacity and minimal leakage of Ni2+ ions.
- As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
- Available in convenient prepacked HisPrep and HisTrap column formats.
- Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
- The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
- Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
- As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Specifications
0.1M HCl, 0.1M NaOH, 8M Urea | |
9.84 in. | |
600cm/hr. | |
3 to 12 | |
6% Cross-linked Agarose | |
Ni-Sepharose |
Ni Sepharose 6 FF chromatography media | |
250mm | |
1 bar | |
90μm | |
4° to 30°C |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Flammable liquid Category 3