missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ni Sepharose™ excel
Ni Sepharose excel affinity media for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).
£414.00 - £5831.83
Specifications
Buffer | 0.01M HCl and 0.01M NaOH (one week), 10mM EDTA, 5mM DTT, 1M NaOH, 5mM TCEP, 20mM beta-Mercapto ethanol and 6M Guanidine-HCl (24 Hours), 500mM Imidazole and 100mM EDTA (2 Hours), 30% 2-Propanol |
---|---|
Product Type | Ni Sepharose excel chromato graphy media |
Flow Rate | 150 to 600cm/hr. |
pH Range | 2 to 12 |
Particle Size | 90μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13415187
|
Cytiva
17371201 |
25 mL |
£414.00
25mL |
In Stock
Log in to see stock. |
|||||
12664557
|
Cytiva
17-3712-02 |
100 mL |
£1521.00
100mL |
In Stock
Log in to see stock. |
|||||
12621206
|
Cytiva
17-3712-03 |
500 mL |
£5831.83
500mL |
In Stock
Log in to see stock. |
|||||
Description
Ni Sepharose excel enables direct loading of samples without having to perform extensive and time-consuming pretreatment procedures.
- Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity.
- Increase target protein yield and decrease degradation through reduced and simplified sample handling.
- Choose between several formats for screening and preparative purification of histidine-tagged proteins.
- Ni Sepharose excel is an IMAC medium precharged with nickel ions very strongly bound to a chelating ligand, allowing loading of samples that usually cause stripping of metal ions.
- Ni Sepharose excel is designed primarily for capture and purification of histidine-tagged proteins secreted into cell culture supernatants from eukaryotic cells such as insect cells or CHO cells.
Specifications
0.01M HCl and 0.01M NaOH (one week), 10mM EDTA, 5mM DTT, 1M NaOH, 5mM TCEP, 20mM beta-Mercapto ethanol and 6M Guanidine-HCl (24 Hours), 500mM Imidazole and 100mM EDTA (2 Hours), 30% 2-Propanol | |
150 to 600cm/hr. | |
90μm | |
4° to 30°C |
Ni Sepharose excel chromato graphy media | |
2 to 12 | |
6% Highly Cross-linked Agarose | |
Ni-Sepharose |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ||Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains Nickel. May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.