missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals Product Code.: 10256410

Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10256410
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10256410

Brand: Thermo Scientific Acros 211085000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-97-0, 7786-81-4
Molecular Formula NiO46H2O
Molecular Weight (g/mol) 262.85
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928Show MoreShow Less
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Melting Point 53.0°C
CAS Min % 98.5
CAS Max % 100.0
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula NiSO4·6H2O
Merck Index 15,6602
Solubility Information Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol
Formula Weight 262.85
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Nickel(II) sulfate hexahydrate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467145 Certificate TypeCertificate of Analysis Date28/10/2024 Catalog Number211080025,211081000,211080100,211085000
Lot NumberA0465410 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number211080025,211081000,211080100,211085000
Lot NumberA0461345 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number211080025,211081000,211080100,211085000
Lot NumberA0459904 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog Number211080025,211081000,211080100,211085000
Lot NumberA0456845 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number211080025,211081000,211080100,211085000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) sulfate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.