missing translation for 'onlineSavingsMsg'
Learn More
Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals Product Code.: 11433870

Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals

Product Code. 11433870
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11433870

Brand: Thermo Scientific Alfa Aesar 089938.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel molybdenum oxide is used as catalyst in petroleum industry for the oil fraction like naptha, kerosene and diesel. It is also used extensively as a chemical intermediate for catalytic purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14177-55-0
Molecular Formula MoNiO4
Molecular Weight (g/mol) 218.639
MDL Number MFCD00016255
InChI Key NLPVCCRZRNXTLT-UHFFFAOYSA-N
Synonym molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+Show MoreShow Less
PubChem CID 84241
IUPAC Name dioxido(dioxo)molybdenum;nickel(2+)
SMILES [O-][Mo](=O)(=O)[O-].[Ni+2]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water (4652mg/L at 20C).
Formula Weight 218.65
Percent Purity 98%
Odor Odorless
Chemical Name or Material Nickel molybdenum oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14K008 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number089938.14,089938.22
Lot NumberY16J008 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number089938.14,089938.22
Lot NumberS01J006 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number089938.22
Lot NumberT10J017 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number089938.14,089938,089938.22
Lot NumberT10J024 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number089938.14,089938,089938.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.