missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Nickel molybdenum oxide is used as catalyst in petroleum industry for the oil fraction like naptha, kerosene and diesel. It is also used extensively as a chemical intermediate for catalytic purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Soluble in water (4652mg/L at 20C). |
Formula Weight | 218.65 |
Percent Purity | 98% |
Odor | Odorless |
Chemical Name or Material | Nickel molybdenum oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'