Learn More
Nickel sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Description
The accumulation of radiation defects in the crystal lattice of nickel in the course of irradiation leads to an increase in the metal sputtering yield from the surface of grains, making it comparable with the sputtering yield from the intergranular region. These result in development of the heavy ion acceleration facilities and for the high-energy heavy-ion doping of deep layers in semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 Ea. |
CAS | 7440-02-0 |
Odor | Odorless |
Molecular Formula | Ni |
MDL Number | MFCD00011137 MFCD06798735 |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
Solubility Information | Insoluble in water. |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
SMILES | [Ni] |
IUPAC Name | nickel |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.