missing translation for 'onlineSavingsMsg'
Learn More
Nitronium tetrafluoroborate, 96%, Thermo Scientific Chemicals Product Code.: 11499586

Nitronium tetrafluoroborate, 96%, Thermo Scientific Chemicals

Product Code. 11499586
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11499586

Brand: Thermo Scientific Alfa Aesar B20167.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolaneShow MoreShow Less
PubChem CID 11073463
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point ∽240°C (decomposition)
Quantity 100 g
Linear Formula NO2BF4
UN Number UN3260
Solubility Information Decomposes in water.
IUPAC Name nitronium;tetrafluoroborate
Formula Weight 132.81
Percent Purity 96%
Sensitivity Moisture sensitive
Chemical Name or Material Nitronium tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251568 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB20167.14,B20167.22,B20167.06
Lot Number10244688 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog NumberB20167.14,B20167.22,B20167.06
Lot Number10238709 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB20167.14,B20167.22,B20167.06
Lot Number10227008 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20167.14,B20167.22,B20167.06
Lot NumberJ23R053 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20167.14,B20167.22,B20167.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitronium tetrafluoroborate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.