missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ NKTEST™ (CE/IVD)
For quantitative determination of the cytotoxic activity of human natural killer (NK) cells.
£583.70 - £2433.90
Specifications
Content And Storage | Reagent A (complete medium) and reagent C (DNA staining solution) must be stored at 2-8°C (in refrigerator). Store Reagent B (Interleukin-2) at >-15°C. The K562 target cells have to be stored at - 70°C |
---|---|
Detection Method | Flow Cytometry, Stain |
Stability | Cells are stable for 12 weeks showing a viability >90%. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15941240
|
BD Biosciences™
660672 |
20 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15991230
|
BD Biosciences™
659179 |
100 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- This reagent kit allows the quantitative determination of the cytotoxic activity of human natural killer (NK) cells.
- Contains cryopreserved prestained K562 target cells, complete medium and a DNA-staining reagent.
- The K562 target cells are labelled with a lipohilic green fluorescent membrane dye in order to discriminate effector and target cells.
- After the incubation period in the cytotoxicity assay, killed target cells are identified by a DNA-stain, which penetrates the dead cells and specifically stains their nuclei.
- In this manner, the percentage of target cells killed by effector NK cells can be determined.
Specifications
Reagent A (complete medium) and reagent C (DNA staining solution) must be stored at 2-8°C (in refrigerator). Store Reagent B (Interleukin-2) at >-15°C. The K562 target cells have to be stored at - 70°C | |
Flow Cytometry, Stain | |
Cells are stable for 12 weeks showing a viability >90%. |