Learn More
Masterflex™ L/S™ Norprene™ High-Pressure Pump Tubing
Designed to pump at higher pressures
£158.00 - £197.00
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11884242
|
Masterflex™
06504-15 |
15 |
£197.00
Each |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
11874242
|
Masterflex™
06504-16 |
16 |
£158.00
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
Specifications
Autoclavable | |
Norprene |