Learn More
Invitrogen™ iBlot™ Transfer Stack, PVDF, mini

Used to transfer proteins with the iBlot Gel Transfer Device
£174.00 - £360.00
Specifications
Length (Metric) | 8 cm |
---|---|
Width (Metric) | 8 cm |
Format | Transfer Stack |
Material | PVDF |
Pore Size | 0.2 μm |
Includes: One box of iBlot Transfer Stacks containing 10 stacks
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
11330982
![]() |
Invitrogen™
IB401032 |
Mini, 8 cm x 8 cm |
£360.00
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11524906
![]() |
Invitrogen™
IB401002 |
Mini, 8 cm x 8 cm |
£174.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The iBlot Transfer Stacks are used to transfer proteins using the iBlot Gel Transfer Device. The iBlot Transfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.
PVDF membrane
- 0.2μm pore size, low fluorescence
- Has higher binding capacity than nitrocellulose
- Preactivated and ready for use without any pretreatment with alcohols
- Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
- Bind proteins due to hydrophobic interactions
- Protein binding capacity: 240μg/cm2
Transfer stack sizes
- The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
- The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room temperature
Specifications
8 cm | |
Transfer Stack | |
0.2 μm | |
iBlot |
8 cm | |
PVDF | |
Mini, 8 cm x 8 cm | |
Sustainable disposal |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.