missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternBreeze™ Wash Solution (16X)

Optimized solution of concentrated buffered saline containing detergent

Brand:  Invitrogen™ WB7003

Product Code. 11590886

  • £96.75 / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 2 x 100mL Antibody Wash (16X)
This item is not returnable. View return policy

Description

Description

Minimizes background and non-specific binding on nitrocellulose (NC) and polyvinylidene difluoride (PVDF) membranes. Sufficient reagents for 20 mini-blots.

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wash Solution
2 x 100 ml Antibody Wash (16X)

Store at +4°C.
WesternBreeze™
Blotting Buffers
Nitrocellulose, PVDF
2 x 100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ WesternBreeze™ Wash Solution (16X) > 2 x 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.