Learn More
Thermo Scientific™ NP-40 Surfact-Amps™ Detergent Solution
Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.
£139.00 - £900.00
Specifications
Form | Liquid |
---|---|
Product Line | Surfact-Amps™ |
Reagent Type | Detergent Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13434269
|
Thermo Scientific™
85124 |
50 mL |
£139.00
50mL |
In Stock
Log in to see stock. |
|||||
11596671
|
Thermo Scientific™
28324 |
6 x 10 mL |
£165.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
13444269
|
Thermo Scientific™
85125 |
500 mL |
£900.00
500mL |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ NP-40 Detergent Surfact-Amps Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.
This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.
Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
NP-40 – a nonionic detergent for use in various protein methods
Accurate – precise 10% detergent solution in ultrapure water
Easy-to-use – solution is simple to dispense and dilute for use
Exceptionally pure – less than 1.0μeq/mL peroxides and carbonyls
Stable – packed in glass ampules under inert nitrogen gas
Detergent Properties:
- Molecular Weight: 617
- Detergent Class: Nonionic
- Aggregation Number: 149
- Micelle Molecular Weight: 90,000
- Critical Micelle Concentration (CMC): 0.29mM (0.0179%, w/v)
- Cloud Point: 80°C
- Dialyzable: No
Specifications
Liquid | |
Surfact-Amps™ | |
Detergent Solution |
For Research Use Only. Not for use in diagnostic procedures.