Learn More
Macherey-Nagel™ Nucleosil™ EC Guard Columns
Brand:  Macherey-Nagel™ 721070.30
 View more versions of this product
Product Code. 12738072
  • £136.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
11%
5 μm
120Å
3 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes
Yes
2 to 8
4 mm
EC Guard Column
NUCLEOSIL™ C18
Acetonitrile/Water
Octadecyl
RP chromatography
Spherical
Hydrophobic
L1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Nucleosil™ EC Guard Columns > Pore Size: 120A