missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA/Protein Column

Isolate RNA and protein from undivided samples. Macherey Nagel™ NucleoSpin™ RNA/Protein Column Set solves the problem of extracting both protein and RNA from the same sample in parallel

Brand:  Macherey-Nagel™ 740933.10

68.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 12718592

  • £84.40 / 10 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • Parallel isolation of RNA and protein from undivided samples
  • No splitting of samples required – reliable interpretation of RNA and protein extracted 
  •  from the same sample (direct correlation)
  • High RNA yield and integrity
  • High protein yield independent of protein size, localization, modification, etc.
  • Easy protein quantification using the MACHEREY-NAGEL Protein Quantification Assay
  • Complete mini kit with NucleoSpin™ Filters (shredders) and recombinant DNase for 
  • DNA-free RNA of high quality
Applications
  • Rapid purification of total RNA and protein from cultured cells and tissue
  • Gene expression profiling, siRNA experiments, analysis of transgenic organisms, drug screening
  • Broad spectrum of starting material tested and proteins already detected
Specifications

Specifications

NucleoSpin™ RNA/Protein Column Set
2mL Collection Tubes, 1.5mL Collection Tubes, NucleoSpin™ Filters, Buffers, RNase-free and rDNase
Spin Column
1Set
10rxns
For Rapid Purification of Total RNA and Protein from Cultured Cells and Tissue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate,ethanol(diluted< 20 %)/dodecyl sulfate, sodium salt/tris-(2-carboxyethyl)ph

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers