missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA/Protein Column
Isolate RNA and protein from undivided samples. Macherey Nagel™ NucleoSpin™ RNA/Protein Column Set solves the problem of extracting both protein and RNA from the same sample in parallel
£84.40 - £3205.00
Specifications
Product Type | NucleoSpin™ RNA/Protein Column Set |
---|---|
Quantity | 1Set |
Includes | 2mL Collection Tubes, 1.5mL Collection Tubes, NucleoSpin™ Filters, Buffers, RNase-free and rDNase |
For Use With (Application) | For Rapid Purification of Total RNA and Protein from Cultured Cells and Tissue |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
12718592
|
Macherey-Nagel™
740933.10 |
10rxns |
£84.40
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12728592
|
Macherey-Nagel™
740933.250 |
250rxns |
£3205.00
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11912382
|
Macherey-Nagel™
740933.50 |
50rxns |
£793.00
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features
- Parallel isolation of RNA and protein from undivided samples
- No splitting of samples required – reliable interpretation of RNA and protein extracted from the same sample (direct correlation)
- High RNA yield and integrity
- High protein yield independent of protein size, localization, modification, etc.
- Easy protein quantification using the MACHEREY-NAGEL Protein Quantification Assay
- Complete mini kit with NucleoSpin™ Filters (shredders) and recombinant DNase for
- DNA-free RNA of high quality
Applications
- Rapid purification of total RNA and protein from cultured cells and tissue
- Gene expression profiling, siRNA experiments, analysis of transgenic organisms, drug screening
- Broad spectrum of starting material tested and proteins already detected
Specifications
NucleoSpin™ RNA/Protein Column Set | |
1Set | |
2mL Collection Tubes, 1.5mL Collection Tubes, NucleoSpin™ Filters, Buffers, RNase-free and rDNase | |
For Rapid Purification of Total RNA and Protein from Cultured Cells and Tissue |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate,ethanol(diluted< 20 %)/dodecyl sulfate, sodium salt/tris-(2-carboxyethyl)ph