missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Soil, Mini kit for DNA from soil
Isolate DNA from soil. Macherey Nagel™ NucleoSpin™ enables soil Isolation of total DNA from diverse soil types
£119.00 - £2610.00
Specifications
Format | Mini Spin Column |
---|---|
Isolation Technology | Silica Membrane Technology |
For Use With (Application) | For Total DNA From Microorganisms in Soil and Sediment |
Sample Volume | <500 mg |
Test Time | 90min. |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12979884
|
Macherey-Nagel™
740780.10 |
10 Preps |
£119.00
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11962402
|
Macherey-Nagel™
740780.50 |
50 Preps |
£570.00
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12718562
|
Macherey-Nagel™
740780.250 |
250 |
£2610.00
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features- Two alternative lysis buffers and a special additive (Enhancer SX) for optimal processing of various soil samples – high yield and high purity
- NucleoSpin™ Bead Tubes with ceramic beads for most efficient cell disruption of Gram-positive and Gram-negative bacteria, archaea, yeast, fungi, and algae in soil, sludge,or sediment
- NucleoSpin™ Inhibitor Removal Column to eliminate all PCR inhibitors, such as humic substances – DNA is ready-to-use for PCR without dilution
- Total DNA from microorganisms in soil and sediment
- Quantify and detect microorganisms from environmental samples
- Typical downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications
Mini Spin Column | |
For Total DNA From Microorganisms in Soil and Sediment | |
90min. |
Silica Membrane Technology | |
<500 mg | |
2 to 10μg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate; guanidine hydrochloride, 2-propanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ NucleoSpin™ Soil, Mini kit for DNA from soil