Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate Product Code.: 10162881
In Stock
to see stock.
Add to basket
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Product Code. 10162881
384, 10 Microplates, 30 Microplates
Click to view available options
No. of Wells:
96
384
No. per Pack:
1 Microplates
10 Microplates
25 Microplates
No. per Case:
100 Microplates
30 Microplates
Product Code. 10162881
Brand: Thermo Scientific™ 242757
In Stock
to see stock.
Add to basket
Add to basket
Color: Clear; Sterility: Sterile; Lid: With Lid
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available
  • Flexible working volumes; 10-100μL
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid With Lid
Culture Area 0.084 cm2/well
Description Non-Treated; w/ lid; Sterile; 10/30
Array 16 x 24
Packaging 10 Pk., 30 Cases
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 30 Microplates
No. per Pack 10 Microplates
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184770 Certificate TypeCertificate of Irradiation Date20/01/2025 Catalog Number242757
Lot Number184770 Certificate TypeCertificate of Compliance Date04/01/2025 Catalog Number242757
Lot Number1326977 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number242757
Lot Number1417524 Certificate TypeCertificate of Processing Date06/12/2024 Catalog Number242757
Lot Number1406155 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number242757
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate >